MilliporeSigma
  • Ordered and ultrathin reduced graphene oxide LB films as hole injection layers for organic light-emitting diode.
Ordered and ultrathin reduced graphene oxide LB films as hole injection layers for organic light-emitting diode.
Nanoscale research letters (2014-10-10)
Yajie Yang, Xiaojie Yang, Wenyao Yang, Shibin Li, Jianhua Xu, Yadong Jiang
ABSTRACT
In this paper, we demonstrated the utilization of reduced graphene oxide (RGO) Langmuir-Blodgett (LB) films as high performance hole injection layer in organic light-emitting diode (OLED). By using LB technique, the well-ordered and thickness-controlled RGO sheets are incorporated between the organic active layer and the transparent conducting indium tin oxide (ITO), leading to an increase of recombination between electrons and holes. Due to the dramatic increase of hole carrier injection efficiency in RGO LB layer, the device luminance performance is greatly enhanced comparable to devices fabricated with spin-coating RGO and a commercial conducting polymer PEDOT:PSS as the hole transport layer. Furthermore, our results indicate that RGO LB films could be an excellent alternative to commercial PEDOT:PSS as the effective hole transport and electron blocking layer in light-emitting diode devices.