MilliporeSigma
  • Anti-leishmanial and anti-trypanosomal potential of polygodial isolated from stem barks of Drimys brasiliensis Miers (Winteraceae).
Anti-leishmanial and anti-trypanosomal potential of polygodial isolated from stem barks of Drimys brasiliensis Miers (Winteraceae).
Parasitology research (2011-01-19)
Daniela S Corrêa, André G Tempone, Juliana Q Reimão, Noemi N Taniwaki, Paulete Romoff, Oriana A Fávero, Patricia Sartorelli, Murilo C Mecchi, João Henrique G Lago
ABSTRACT
Parasitic protozoan diseases affect the poorest population in developing countries. Leishmaniasis and Chagas disease have been included among the most important threats for public health in Central and South American continent, with few therapeutic alternatives and highly toxic drugs. In the course of selection of novel drug candidates, we studied the anti-protozoal potential of Drimys brasiliensis. Thus, the crude hexane extract from stem bark as well as its main derivative, the sesquiterpene polygodial, were tested using in vitro assays. The crude hexane extract and polygodial showed activity against Leishmania spp. in the range between 22 and 62 μg/mL, but polygodial demonstrated high parasite selectivity towards Trypanosoma cruzi trypomastigotes (2 μg/mL), with a selectivity index of 19. Finally, polygodial showed a leishmanicidal effect, inducing intense ultrastructural damages in Leishmania in short-time incubation. The obtained results suggested that polygodial could be used as a tool for drug design studies against protozoan diseases and as a candidate molecule for further in vivo studies against T. cruzi.
Sigma-Aldrich
Polygodial, ≥97% (HPLC)