MilliporeSigma
  • Troponin I and Tropomyosin regulate chromosomal stability and cell polarity.
Troponin I and Tropomyosin regulate chromosomal stability and cell polarity.
Journal of cell science (2009-07-02)
Virender Kumar Sahota, Benjamin Filip Grau, Alicia Mansilla, Alberto Ferrús
ABSTRACT
The Troponin-Tropomyosin (Tn-Tm) complex regulates muscle contraction through a series of Ca(2+)-dependent conformational changes that control actin-myosin interactions. Members of this complex in Drosophila include the actin-binding protein Troponin I (TnI), and two Tropomyosins (Tm1 and Tm2), which are thought to form heterodimers. We show here that pre-cellular embryos of TnI, Tm1 and Tm2 mutants exhibit abnormal nuclear divisions with frequent loss of chromosome fragments. During cellularization, apico-basal polarity is also disrupted as revealed by the defective location of Discs large (Dlg) and its ligand Rapsynoid (Raps; also known as Partner of Inscuteable, Pins). In agreement with these phenotypes in early development, on the basis of RT-PCR assays of unfertilized eggs and germ line mosaics of TnI mutants, we also show that TnI is part of the maternal deposit during oogenesis. In cultures of the S2 cell line, native TnI is immunodetected within the nucleus and immunoprecipitated from nuclear extracts. SUMOylation at an identified site is required for the nuclear translocation. These data illustrate, for the first time, a role for TnI in the nucleus and/or the cytoskeleton of non-muscle cells. We propose that the Tn-Tm complex plays a novel function as regulator of motor systems required to maintain nuclear integrity and apico-basal polarity during early Drosophila embryogenesis.