MilliporeSigma
  • A helix-loop-helix protein related to the immunoglobulin E box-binding proteins.
A helix-loop-helix protein related to the immunoglobulin E box-binding proteins.
Molecular and cellular biology (1990-08-01)
C S Carr, P A Sharp
ABSTRACT
A human cDNA encoding a novel protein in the helix-loop-helix family has been isolated by screening a bacteriophage expression library with a probe containing the binding site for major late transcription factor. The protein encoded by this cDNA, TFEB, probably recognizes E-box sequences in the heavy-chain immunoglobulin enhancer.