MilliporeSigma
  • Synthesis, biochemical and molecular modelling studies of antiproliferative azetidinones causing microtubule disruption and mitotic catastrophe.
Synthesis, biochemical and molecular modelling studies of antiproliferative azetidinones causing microtubule disruption and mitotic catastrophe.
European journal of medicinal chemistry (2011-08-16)
Niamh M O'Boyle, Miriam Carr, Lisa M Greene, Niall O Keely, Andrew J S Knox, Thomas McCabe, David G Lloyd, Daniela M Zisterer, Mary J Meegan
ABSTRACT
The structure-activity relationships of antiproliferative β-lactams, focusing on modifications at the 4-position of the β-lactam ring, is described. Synthesis of this series of compounds was achieved utilizing the Staudinger and Reformatsky reactions. The antiproliferative activity was assessed in MCF-7 cells, where the 4-(4-ethoxy)phenyl substituted compound 26 displayed the most potent activity with an IC(50) value of 0.22 μM. The mechanism of action was demonstrated to be by inhibition of tubulin polymerisation. Cell exposure to combretastatin A-4 and 26 led to arrest of MCF-7 cells in the G2/M phase of the cell cycle and induction of apoptosis. Additionally, mitotic catastrophe for combretastatin A-4 and for 26 was demonstrated in breast cancer cells for the first time, as evidenced by the formation of giant, multinucleated cells.
Sigma-Aldrich
Azetidine, 98%