MilliporeSigma
  • Perfluorononanoic acid-induced apoptosis in rat spleen involves oxidative stress and the activation of caspase-independent death pathway.
Perfluorononanoic acid-induced apoptosis in rat spleen involves oxidative stress and the activation of caspase-independent death pathway.
Toxicology (2009-11-03)
Xuemei Fang, Yixing Feng, Jianshe Wang, Jiayin Dai
ABSTRACT
Perfluoroalkyl acid (PFAA)-induced apoptosis has been reported in many cell types. However, minimal information on its mode of action is available. This study explored the possible involvement of apoptotic signaling pathways in a nine-carbon-chain length PFAA-perfluorononanoic acid (PFNA)-induced splenocyte apoptosis. After a 14-day exposure to PFNA, rat spleens showed dose-dependent levels of apoptosis. The production of pro-inflammatory and anti-inflammatory cytokines was significantly increased and decreased, respectively. However, protein levels of tumor necrosis factor receptor 1 (TNFR1), fas-associated protein with death domain (FADD), caspase 8 and caspase 3, which are involved in inflammation-related and caspase-dependent apoptosis, were discordant. Peroxisome proliferator-activated receptors alpha (PPARalpha) and PPARgamma genes expression was up-regulated in rats treated with 3 or 5 mg/kg/day of PFNA, and the level of hydrogen peroxide (H2O2) increased concurrently in rats treated with the highest dose. Moreover, superoxide dismutase (SOD) activity and Bcl-2 protein levels were dramatically decreased in spleens after treatment with 3 and 5 mg/kg/day of PFNA. However, protein levels of Bax were unchanged. Apoptosis-inducing factor (AIF), an initiator of caspase-independent apoptosis, was significantly increased in all PFNA-dosed rats. Thus, oxidative stress and the activation of a caspase-independent apoptotic signaling pathway contributed to PFNA-induced apoptosis in rat splenocytes.
Sigma-Aldrich
Perfluorononanoic acid, 97%