MilliporeSigma
  • Quantitative determination of erythromycylamine in human plasma by liquid chromatography-mass spectrometry and its application in a bioequivalence study of dirithromycin.
Quantitative determination of erythromycylamine in human plasma by liquid chromatography-mass spectrometry and its application in a bioequivalence study of dirithromycin.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-02-26)
Yun-Qing Liu, Qi-Yuan Chen, Ben-Mei Chen, Shao-Gang Liu, Fu-Liang Deng, Ping Zhou
ABSTRACT
A sensitive, rapid liquid chromatographic-electrospray ionization mass spectrometric method for determination of erythromycylamine in human plasma was developed and validated. Erythromycylamine in plasma (0.2 mL) was extracted with ethyl acetate, the organic phase was transferred to another clear 1.5 mL Eppendorf tube and evaporated to dryness under gentle nitrogen stream at 45 degrees C, and the residue was dissolved in 100 microL of mobile phase. The samples were separated using a Thermo Hypersil HyPURITY C18 reversed-phase column (150 mm x 2.1 mm I.D., 5 microm). A mobile phase containing 10 mM of ammonium acetate (pH = 6.4)-acetonitrile-methanol (50:10:40, v/v/v) was used isocratically eluting at a flow rate of 0.2 mL/min. Erythromycylamine and its internal standard (IS), midecamycin, were measured by electrospray ion source in positive selective ion monitoring mode. The method demonstrated that good linearity ranged from 4.5 to 720 ng/mL with r = 0.9997. The limit of quantification for erythromycylamine in plasma was 4.5 ng/mL with good accuracy and precision. The mean extraction recovery of the method was higher than 75.1% and 72.7% for erythromycylamine and IS, respectively. The intra-day and inter-day precision ranged from 5.2% to 6.4% and 5.6-9.3% (relative standard deviation, RSD), respectively. The established method has been successfully applied to a bioequivalence study of two dirithromycin formulations for 18 healthy volunteers.