MilliporeSigma
  • A comparison of two intramuscular doses of xylazine-ketamine combination and tolazoline reversal in llamas.
A comparison of two intramuscular doses of xylazine-ketamine combination and tolazoline reversal in llamas.
Veterinary anaesthesia and analgesia (2004-04-01)
William R DuBois, Tulio M Prado, Jeff C H Ko, Ronald E Mandsager, Gregor L Morgan
ABSTRACT
To evaluate the anesthetic and cardiorespiratory effects of two doses of intramuscular xylazine/ketamine in llamas, and to determine if an intramuscular injection of tolazoline would shorten the anesthesia recovery time. Prospective randomized study. Six castrated male llamas. Each llama received a low dose (LD) (0.4 mg kg(-1) xylazine and 4 mg kg(-1) ketamine) and high dose (HD) (0.8 mg kg(-1) xylazine and 8 mg kg(-1) ketamine). Time to sedation, duration of lateral recumbency and analgesia, pulse, respiratory rate, hemoglobin oxygen saturation, arterial blood pressure, blood gases, and the electrocardiogram were monitored and recorded during anesthesia. Three llamas in each treatment were randomized to receive intramuscular tolazoline (2 mg kg(-1)) after 30 minutes of lateral recumbency. Onset of sedation, lateral recumbency, and analgesia was rapid with both treatments. The HD was able to provide at least 30 minutes of anesthesia in all six llamas. The LD provided only 30 minutes of anesthesia in two out of six llamas. Respiratory depression and hypoxemia were seen in the HD treatment during the first 10 minutes of lateral recumbency. Two llamas were severely hypoxemic during this period and were given nasal oxygen for five minutes. Heart rate decreased, but there were no significant changes in blood pressure. Tolazoline significantly shortened the duration of recumbency in the HD treatment. The HD provided more consistent clinical effects in llamas than did the LD. Intramuscular tolazoline shortens the duration of lateral recumbency in llamas anesthetized with this combination. Both doses appear to be very effective in providing restraint in llamas. The LD may be used for procedures requiring a short period of anesthesia or restraint. The HD could be used when a longer duration of anesthesia is desired. Supplemental oxygen should be available if using the HD. Tolazoline (IM) shortened the recovery time with this combination in llamas.