MilliporeSigma
  • Cell-based evidence for aminopeptidase N/CD13 inhibitor actinonin targeting of MT1-MMP-mediated proMMP-2 activation.
Cell-based evidence for aminopeptidase N/CD13 inhibitor actinonin targeting of MT1-MMP-mediated proMMP-2 activation.
Cancer letters (2009-03-07)
Asmaa Sina, Simon Lord-Dufour, Borhane Annabi
ABSTRACT
Recent profiling has identified the aminopeptidase N/CD13 inhibitor actinonin as a selective soluble secreted matrix metalloproteinase (MMP) inhibitor. Given that actinonin's effects against membrane-bound MMPs remain unknown and that MT1-MMP has been linked to chemo- and radio-therapy resistance in brain tumor development, we therefore assessed MT1-MMP functional inhibition by actinonin in U87 glioblastoma cells. We show that actinonin inhibits concanavalin-A (ConA)-induced proMMP-2 activation, while it does not inhibit ConA-induced MT1-MMP gene expression suggesting post-transcriptional effects of the drug possibly mediated through the membrane-anchored protease regulator RECK. Specific gene silencing of MT1-MMP with siRNA abrogated the ability of ConA to activate proMMP-2. Functional recombinant MT1-MMP whose constitutive expression led to proMMP-2 activation was also efficiently antagonized by actinonin. We provide evidence for actinonin's new therapeutic application in the direct targeting of MT1-MMP-mediated proMMP-2 activation, an essential step in both brain tumor infiltration and in brain tumor-associated angiogenesis.
Sigma-Aldrich
Actinonin