MilliporeSigma
  • [Effects of diphenyl derivatives on electrophoretic mobility of murine T lymphocytes].
[Effects of diphenyl derivatives on electrophoretic mobility of murine T lymphocytes].
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994) (2013-08-21)
O V Dolga, N Kh Pogoriela, O S Bohorad-Kobel's'ka, N M Zholobak, S O Zanoza, S A Liakhov, I S Mahura
ABSTRACT
The early changes of electrophoretic mobility (EPM) of murine T lymphocytes induced by structural analogues of amixine-dihydrochloryde 4,4'-bis-[2(diethylamino)ethoxy]diphenyl (compound 1) and dihydrochloryde 2-methoxycarbonil-4,4'-bis-[2(diethylamino)ethoxy]diphenyl (compound 2) were studied by electrophoresis technique. During the interval 0-2 hours all compounds increased the absolute values of EPM in comparison with control. These changes were of the same kind--distinctions were quantitative. Amixine and compound 1 during the interval 2-4 hours additionally increased the EPM. The compound 2, on the contrary, decreased the EPM. It was shown that the opposite effects of the aforementioned compounds were caused by the fact that amixine and compound 1 induce, and compound 2 does not induce IFN production in T lymphocytes in vitro. The results of our experiments are important for understanding of the mechanisms of immunomodulating effect of amixine and its structural analogues.