MilliporeSigma
  • Pharmacokinetic evaluation of olopatadine for the treatment of allergic rhinitis and conjunctivitis.
Pharmacokinetic evaluation of olopatadine for the treatment of allergic rhinitis and conjunctivitis.
Expert opinion on drug metabolism & toxicology (2011-10-29)
Andrew S Nickels, Vesselin Dimov, Raoul Wolf
ABSTRACT
Olopatadine hydrochloride is an antihistamine and mast cell stabilizer available in three forms, including oral, intranasal and ocular preparations. Most of the practical applications focus on the use of olopatadine for the treatment of allergic rhinitis and conjunctivitis via intranasal and ocular routes. This article was formed from a comprehensive literature search with information taken from meta-analyses, systematic reviews, treatment guidelines and clinical studies on children and adults. Articles that have been selected evaluate the use of intranasal and ocular antihistamines and their role in allergic rhinitis and conjunctivitis. Olopatadine is significantly more effective than placebos in alleviating the symptoms of allergic rhinitis and conjunctivitis. Olopatadine is a viable alternative and addition to the mainstay therapy of these conditions with intranasal steroids and oral antihistamines. The compliance of the patients would be improved if a once-per-day formulation of olopatadine was developed for intranasal application. The future treatments of allergic rhinitis will probably involve a combination of intranasal antihistamine and steroid because clinical trials have demonstrated an improved efficacy without a significant increase in adverse effects.
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)