MilliporeSigma
  • Efficacy of eprinomectin pour-on against Dictyocaulus arnfieldi infection in donkeys (Equus asinus).
Efficacy of eprinomectin pour-on against Dictyocaulus arnfieldi infection in donkeys (Equus asinus).
Veterinary journal (London, England : 1997) (2011-01-11)
Vincenzo Veneziano, Antonio Di Loria, Roberto Masucci, Rossella Di Palo, Emanuele Brianti, Cengiz Gokbulut
ABSTRACT
A trial to assess the efficacy of eprinomectin (EPM) against the lungworm Dictyocaulus arnfieldi was carried out on 15, naturally-infected donkeys. Ten animals were treated with a 'pour-on' EPM preparation (at a dose of 0.5 mg/kg bodyweight), and five animals acted as controls. Faecal larval counts were carried out two days before treatment, on day of treatment and 7, 14, 21 and 28 days post-treatment with the anthelmintic. EPM was 100% effective in eliminating faecal larvae from day 7, until the end of study at day 28. No adverse drug-reactions or side-effects were observed in any of the treated donkeys.
Product Description
Supelco
Eprinomectin, PESTANAL®, analytical standard, mixture of isomers