MilliporeSigma
  • Potentiometric sensing of butyrylcholinesterase based on in situ generation and detection of substrates.
Potentiometric sensing of butyrylcholinesterase based on in situ generation and detection of substrates.
Chemical communications (Cambridge, England) (2009-02-14)
Jiawang Ding, Wei Qin
ABSTRACT
A novel reagentless biosensor has been developed in which the traditional ion selective electrode is used as a controlled-release system for in situ generation and detection of enzyme substrates.
Sigma-Aldrich
Butyrylcholine chloride, ≥98%