MilliporeSigma
  • Comparative study on free amino acid composition of wild edible mushroom species.
Comparative study on free amino acid composition of wild edible mushroom species.
Journal of agricultural and food chemistry (2008-10-24)
Bárbara Ribeiro, Paula B Andrade, Branca M Silva, Paula Baptista, Rosa M Seabra, Patrícia Valentão
ABSTRACT
A comparative study on the amino acid composition of 11 wild edible mushroom species (Suillus bellini, Suillus luteus, Suillus granulatus, Tricholomopsis rutilans, Hygrophorus agathosmus, Amanita rubescens, Russula cyanoxantha, Boletus edulis, Tricholoma equestre, Fistulina hepatica, and Cantharellus cibarius) was developed. To define the qualitative and quantitative profiles, a derivatization procedure with dabsyl chloride was performed, followed by HPLC-UV-vis analysis. Twenty free amino acids (aspartic acid, glutamic acid, asparagine, glutamine, serine, threonine, glycine, alanine, valine, proline, arginine, isoleucine, leucine, tryptophan, phenylalanine, cysteine, ornithine, lysine, histidine, and tyrosine) were determined. B. edulis and T. equestre were revealed to be the most nutritional species, whereas F. hepatica was the poorest. The different species exhibited distinct free amino acid profiles. The quantification of the identified compounds indicated that, in a general way, alanine was the major amino acid. The results show that the analyzed mushroom species possess moderate amino acid contents, which may be relevant from a nutritional point of view because these compounds are indispensable for human health. A combination of different mushroom species in the diet would offer good amounts of amino acids and a great diversity of palatable sensations.
Sigma-Aldrich
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride, ≥97.5% (AT)