MilliporeSigma
  • Novel method to reduce fishy aftertaste in wine and seafood pairing using alcohol-treated yeast cells.
Novel method to reduce fishy aftertaste in wine and seafood pairing using alcohol-treated yeast cells.
Journal of agricultural and food chemistry (2012-05-29)
Toshikazu Tsuji, Keiko Kanai, Aki Yokoyama, Takayuki Tamura, Kenichi Hanamure, Kanako Sasaki, Ryoji Takata, Satoshi Yoshida
ABSTRACT
"Fishy aftertaste" is sometimes perceived in wine consumed with seafood. Iron in wine has been reported to be a key compound that produces fishy aftertaste. However, cost-effective methods to remove iron from wine have not been developed. Here, we describe a cost-effective and safe iron adsorbent consisting of alcohol-treated yeast (ATY) cells based on the observation that nonviable cells adsorbed iron after completion of fermentation. Treatment of cells with more than 40% (v/v) ethanol killed them without compromising their ability to adsorb iron. Drying the ATY cells did not reduce iron adsorption. Use of ATY cells together with phytic acid had a synergistic effect on iron removal. We term this means of removing iron the "ATY-PA" method. Sensory analysis indicated that fishy aftertaste in wine-seafood pairings was not perceived if the wine had been pretreated with both ATY cells and phytic acid.