MilliporeSigma
  • A highly selective turn-on colorimetric, red fluorescent sensor for detecting mobile zinc in living cells.
A highly selective turn-on colorimetric, red fluorescent sensor for detecting mobile zinc in living cells.
Inorganic chemistry (2010-10-30)
Pingwu Du, Stephen J Lippard
ABSTRACT
We describe ZRL1, a turn-on colorimetric and red fluorescent zinc ion sensor. The Zn(2+)-promoted ring opening of the rhodamine spirolactam ring in ZRL1 evokes a 220-fold fluorescence turn-on response. In aqueous media, ZRL1 turn-on luminescence is highly selective for Zn(2+) ions, with no significant response to other competitive cations, including Na(+), K(+), Ca(2+), Mg(2+), Mn(2+), Fe(2+), Co(2+), Ni(2+), Cu(2+), Cd(2+), or Hg(2+). In addition to these characteristics, preliminary results indicate that ZRL1 can be delivered to living cells and can be used to monitor changes in intracellular Zn(2+) levels.