MilliporeSigma
  • Immunoarchitecture of normal human bone marrow: a study of frozen and fixed tissue sections.
Immunoarchitecture of normal human bone marrow: a study of frozen and fixed tissue sections.
Human pathology (1992-06-01)
S S Shin, K Sheibani, J Kezirian, A Nademanee, S J Forman, S K Lee, C D Winberg
ABSTRACT
To provide baseline information on the immunoarchitecture of normal bone marrow, we studied cryostat-cut, frozen, and paraffin-embedded, fixed tissue sections prepared from 21 core biopsies of normal bone marrow obtained during bone marrow harvests for transplantation. A large panel of antibodies was applied that included, for frozen tissue, Leu-6 (CD1), T11 (CD2), Leu-3a (CD4), Leu-1 (CD5), Leu-2a (CD8), J5 (CD10), My7 (CD13), Leu-11 (CD16), B4 (CD19), B1 (CD20), B2 (CD21), Tac (CD25), My9 (CD33), T200 (CD45), NKH-1 (CD56), kappa and lambda chains, beta F1, Ki-67, HLA-DR, TQ1, and keratin, and for fixed tissue, leukocyte common antigen (CD45), L26 (CD20), LN1 (CDw75), LN2 (CD74), LN3, LN4, LN5, MB1 (CD45R), MB2, MT1 (CD43), MT2 (CD45R), UCHL1 (CD45R0), BM1, Ki-1 (CD30), Leu-M1 (CD15), lysozyme, KP1 (CD68), actin, S100, neuron-specific enolase, vimentin, and keratin. On fresh-frozen sections CD19 and CD2 were the most reliable and sensitive markers for B and T cells, staining 5% and 9% of marrow cells, respectively. Immunoglobulins generally showed heavy background staining, which frequently precluded an accurate assessment. The CD4 to CD8 ratio in the bone marrow was reversed from that of peripheral blood. On fixed tissues, leukocyte common antigen was found in 14% of the marrow cells, corresponding roughly to the lymphocyte population. L26, a pan-B-cell marker, stained 3% of the marrow cells. Among the other B-cell markers, LN1 and MB2 stained a large number of cells (40% to 70%), indicating reactivity with cells of the myeloid or erythroid series in addition to lymphocytes. Among the T-cell markers, UCHL1 and MT1 stained 66% and 50% of the cells, respectively, which could be explained by their cross-reactivity with myeloid cells. Nonspecific myelomonocytic markers (Leu-M1, KP1, and lysozyme) also showed reactivity in a high percentage of cells. No particular architectural distribution patterns of B or T lymphocytes were noted in either frozen or fixed bone marrow specimens. The results of this study provide normal baseline data for the immunohistologic application of hematopoietic and lymphoid markers on frozen or fixed bone marrow biopsy specimens.