MilliporeSigma
  • A 25-kD inhibitor of actin polymerization is a low molecular mass heat shock protein.
A 25-kD inhibitor of actin polymerization is a low molecular mass heat shock protein.
The Journal of cell biology (1991-07-01)
T Miron, K Vancompernolle, J Vandekerckhove, M Wilchek, B Geiger
ABSTRACT
The 25-kD inhibitor of actin polymerization (25-kD IAP), isolated from turkey smooth muscle (Miron, T., M. Wilchek, and B. Geiger, 1988. Eur. J. Biochem. 178:543-553), is shown here to be a low molecular mass heat shock protein (HSP). Direct sequence analysis of the purified protein, as well as cloning and sequencing of the respective cDNA, disclosed a high degree of homology (67% identity, 80% similarity) to the human 27-kD HSP. Southern blot of chicken genomic DNA disclosed one band, suggesting the presence of a single gene, and Northern blot analysis revealed abundant transcript of approximately 1 kb in gizzard and heart tissues and lower amounts in total 18-d chick embryo RNA and in cultured fibroblasts. Exposure of the latter cells to 45 degrees C resulted in over 15-fold increase in the apparent level of the 25-kD IAP protein, confirming that its expression is regulated by heat shock. Immunofluorescent microscopic localization indicated that after heat treatment, the levels of the 25-kD IAP were markedly increased and the protein was apparently associated with cytoplasmic granules. Heat shock also had a transient, yet prominent, effect on the microfilament system in cultured fibroblasts: stress fibers disintegrated within 10-15 min after incubation at 45 degrees C, yet upon further incubation at the elevated temperature, conspicuous actin bundles were apparently reformed.