MilliporeSigma
  • 8C-like cells capture the human zygotic genome activation program in vitro.
8C-like cells capture the human zygotic genome activation program in vitro.
Cell stem cell (2022-02-27)
Jasmin Taubenschmid-Stowers, Maria Rostovskaya, Fátima Santos, Sebastian Ljung, Ricard Argelaguet, Felix Krueger, Jennifer Nichols, Wolf Reik
ABSTRACT
The activation of the embryonic genome marks the first major wave of transcription in the developing organism. Zygotic genome activation (ZGA) in mouse 2-cell embryos and 8-cell embryos in humans is crucial for development. Here, we report the discovery of human 8-cell-like cells (8CLCs) among naive embryonic stem cells, which transcriptionally resemble the 8-cell human embryo. They express ZGA markers, including ZSCAN4 and LEUTX, and transposable elements, such as HERVL and MLT2A1. 8CLCs show reduced SOX2 levels and can be identified using TPRX1 and H3.Y marker proteins in vitro. Overexpression of the transcription factor DUX4 and spliceosome inhibition increase human ZGA-like transcription. Excitingly, the 8CLC markers TPRX1 and H3.Y are also expressed in ZGA-stage 8-cell human embryos and may thus be relevant in vivo. 8CLCs provide a unique opportunity to characterize human ZGA-like transcription and might provide critical insights into early events in embryogenesis in humans.
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.