MilliporeSigma
  • Distinct IL-4-induced gene expression, proliferation, and intracellular signaling in germinal center B-cell-like and activated B-cell-like diffuse large-cell lymphomas.
Distinct IL-4-induced gene expression, proliferation, and intracellular signaling in germinal center B-cell-like and activated B-cell-like diffuse large-cell lymphomas.
Blood (2004-12-14)
Xiaoqing Lu, Hovav Nechushtan, Feiying Ding, Manuel F Rosado, Rakesh Singal, Ash A Alizadeh, Izidore S Lossos
ABSTRACT
Diffuse large B-cell lymphomas (DLBCLs) can be subclassified into germinal center B-cell (GCB)-like and activated B-cell (ABC)-like tumors characterized by long and short survival, respectively. In contrast to ABC-like DLBCL, GCB-like tumors exhibit high expression of components of the interleukin 4 (IL-4) signaling pathway and of IL-4 target genes such as BCL6 and HGAL, whose high expression independently predicts better survival. These observations suggest distinct activity of the IL-4 signaling pathway in DLBCL subtypes. Herein, we demonstrate similar IL-4 expression but qualitatively different IL-4 effects on GCB-like and ABC-like DLBCL. In GCB-like DLBCL, IL-4 induces expression of its target genes, activates signal transducers and activators of transcription 6 (STAT6) signaling, and increases cell proliferation. In contrast, in the ABC-like DLBCL, IL-4 activates AKT, decreases cell proliferation by cell cycle arrest, and does not induce gene expression due to aberrant Janus kinase (JAK)-STAT6 signaling attributed to STAT6 dephosphorylation. We found distinct expression profiles of tyrosine phosphatases in DLBCL subtypes and identified putative STAT6 tyrosine phosphatases-protein tyrosine phosphatase nonreceptor type 1 (PTPN1) and PTPN2, whose expression is significantly higher in ABC-like DLBCL. These differences in tyrosine phosphatase expression might underlie distinct expression profiles of some of the IL-4 target genes and could contribute to a different clinical outcome of patients with GCB-like and ABC-like DLBCLs.