MilliporeSigma
Millisecond, micron precision multi-whisker detector.
PloS one (2013-09-12)
Stephen K Grady, Thanh T Hoang, Shree Hari Gautam, Woodrow L Shew
ABSTRACT
The neural mechanisms of somatosensory information processing in the rodent vibrissae system are a topic of intense debate and research. Certain hypotheses emphasize the importance of stick-slip whisker motion, high-frequency resonant vibrations, and/or the ability to decode complex textures. Other hypotheses focus on the importance of integrating information from multiple whiskers. Tests of the former require measurements of whisker motion that achieve high spatiotemporal accuracy without altering the mechanical properties of whiskers. Tests of the latter require the ability to monitor the motion of multiple whiskers simultaneously. Here we present a device that achieves both these requirements for two-dimensional whisker motion in the plane perpendicular to the whiskers. Moreover, the system we present is significantly less expensive (<$2.5 k) and simpler to build than alternative devices which achieve similar detection capabilities. Our system is based on two laser diodes and two linear cameras. It attains millisecond temporal precision and micron spatial resolution. We developed automated algorithms for processing the data collected by our device and benchmarked their performance against manual detection by human visual inspection. By this measure, our detection was successful with less than 10 µm deviation between the automated and manual detection, on average. Here, we demonstrate its utility in anesthetized rats by measuring the motion of multiple whiskers in response to an air puff.