MilliporeSigma
  • Inhibition of endoplasmic reticulum stress ameliorates cardiovascular injury in a rat model of metabolic syndrome.
Inhibition of endoplasmic reticulum stress ameliorates cardiovascular injury in a rat model of metabolic syndrome.
Journal of molecular and cellular cardiology (2020-04-21)
Eman Radwan, Marwa H Bakr, Salma Taha, Sally A Sayed, Alshaimaa A Farrag, Maha Ali
ABSTRACT
Metabolic (Met) syndrome is characterized by hypertension, insulin resistance and dyslipidaemia with high risk of cardiovascular disease. Endoplasmic reticulum (ER) stress is a key contributor in the pathogenesis of Met syndrome. The current study investigates the effect of Tauroursodeoxycholate (TUDCA), an ER stress inhibitor, on Met syndrome-induced cardiovascular complications and the possible underlying signalling mechanisms. Met syndrome was induced in rats, which were then treated with TUDCA. Body weight, blood pressure, glucose tolerance and insulin tolerance tests were performed. ER stress, survival and oxidative stress markers were measured in heart and aorta tissue. The results showed that TUDCA improved metabolic parameters in rats with Met syndrome. Treatment mitigated the Met syndrome-induced cardiovascular complications through upregulating survival markers and downregulating ER and oxidative stress markers. These results highlight the protective effect of ER stress inhibition as a potential target in the management of cardiovascular complications associated with Met syndrome.