MilliporeSigma
  • A novel m.11406 T > A mutation in mitochondrial ND4 gene causes MELAS syndrome.
A novel m.11406 T > A mutation in mitochondrial ND4 gene causes MELAS syndrome.
Mitochondrion (2020-07-14)
Yan Lin, Xuebi Xu, Dandan Zhao, Fuchen Liu, Yuebei Luo, Jixiang Du, Dongdong Wang, Kunqian Ji, Yuying Zhao, Chuanzhu Yan
ABSTRACT
Pathogenic point mutations of mitochondrial DNA (mtDNA) are associated with a large number of heterogeneous diseases involving multiple systems with which patients may present with a wide range of clinical phenotypes. In this study, we describe a novel heteroplasmic missense mutation, m.11406 T > A, of the ND4 gene encoding the subunit 4 of mitochondrial complex I in a 32-year-old woman with recurrent epileptic seizure, headache and bilateral hearing loss. Skeletal muscle histochemistry demonstrated that approximately 20% of fibers were cytochrome C oxidase (COX) deficient with increased activity of succinate dehydrogenase (SDH). Further investigations in muscle specimens showed significantly reduced level of ND4 protein. It is interesting that the subunits of complex I (ND1 and NDFUB8) and complex IV(CO1) were also remarkably decreased. These findings indicate that ND1, NDFUB8 and CO1 are more susceptible than other subunits to mutations in the mitochondrial ND4 gene.