MilliporeSigma
  • Low-dose exposure and immunogenicity of transgenic maize expressing the Escherichia coli heat-labile toxin B subunit.
Low-dose exposure and immunogenicity of transgenic maize expressing the Escherichia coli heat-labile toxin B subunit.
Environmental health perspectives (2007-04-14)
April J Beyer, Kan Wang, Amber N Umble, Jeffrey D Wolt, Joan E Cunnick
ABSTRACT
Transgenic maize, which produces the nontoxic B subunit of the Escherichia coli heat-labile toxin (LT-B) in seed, has proven to be an effective oral immunogen in mice. Currently, there is considerable concern over accidental consumption of transgenic maize expressing LT-B by humans and domestic animals. We have yet to define nonimmunogenic levels of transgenic LT-B when ingested. Our goal in this study was to determine the highest dose of LT-B orally administered in mice that does not result in a measurable immune response. We defined an immune response as specific serum or mucosal IgG or IgA significantly greater than background after three feedings (0.0002-20 mug) or a priming response induced by the intermittent feeding. We fed transgenic maize pellets on days 0, 7, 21, and 49 and collected serum and fecal samples weekly. Serum was analyzed for LT-B-specific IgG and IgA, and feces was analyzed for LT-B-specific IgA. We observed a dose-dependent anti-LT-B antibody response with high specific antibody concentrations in groups fed high doses (0.2, 2, 20 mug) of LT-B maize. Mice fed 0.02 mug LT-B demonstrated immune priming in 62.5% of the animals. Mice that were fed </= 0.002 mug LT-B showed no increase in specific antibody nor did they demonstrate immune priming, indicating that 0.002 mug LT-B was the highest nonimmunogenic dose tested. Our results demonstrate that LT-B derived from transgenic maize is immunogenic at nanogram levels when orally administered to mice.
Sigma-Aldrich
Mouse IgG1-k Negative Control, clone MOPC-21, Azide Free Antibody, clone MOPC-21, from mouse