MilliporeSigma
  • Inhibitory Effects of PRG4 on Migration and Proliferation of Human Venous Cells.
Inhibitory Effects of PRG4 on Migration and Proliferation of Human Venous Cells.
The Journal of surgical research (2020-04-23)
Lei Wang, Shinsuke Kikuchi, Tannin A Schmidt, Max Hoofnagle, Thomas N Wight, Nobuyoshi Azuma, Gale L Tang, Michael Sobel, Gautum R Velamoor, Nahush A Mokadam, Richard D Kenagy
ABSTRACT
Proteoglycan 4 (PRG4; lubricin) is a member of two gene co-expression network modules associated with human vein graft failure. However, little is known about PRG4 and the vascular system. Therefore, we have investigated the effects of recombinant human PRG4 (rhPRG4) on cell migration and proliferation in human veins. Effects of rhPRG4 on cell migration, proliferation, and neointima formation were determined in human venous tissue and cultured venous smooth muscle cells (SMCs), adventitial cells, and endothelial cells. Expression of PRG4 by cultured human saphenous veins, failed vein grafts, and varicose veins was determined by immunostaining or Western blotting. Limited expression of PRG4 in fresh saphenous veins was dramatically increased around medial SMCs after culture ex vivo. rhPRG4 inhibited the migration of cultured SMCs, adventitial cells, and endothelial cells, as well as the proliferation of endothelial cells. rhPRG4 also inhibited the migration of SMCs and adventitial cells from tissue explants, but there was no effect on cell proliferation or neointima formation in ex vivo whole veins. Finally, PRG4 was largely absent in two examples of venous pathology, that is, failed human vein grafts and varicose veins. Although rhPRG4 can inhibit the migration of venous SMCs, endothelial cells, and adventitial cells, and the proliferation of endothelial cells, PRG4 was only increased around medial SMCs in veins after ex vivo culture. PRG4 was not observed around medial SMCs in failed human vein grafts and varicose veins, suggesting the possibility that a failure of PRG4 upregulation may promote these pathologies.