MilliporeSigma
  • Triggering a switch from basal- to luminal-like breast cancer subtype by the small-molecule diptoindonesin G via induction of GABARAPL1.
Triggering a switch from basal- to luminal-like breast cancer subtype by the small-molecule diptoindonesin G via induction of GABARAPL1.
Cell death & disease (2020-08-18)
Minmin Fan, Jingwei Chen, Jian Gao, Wenwen Xue, Yixuan Wang, Wuhao Li, Lin Zhou, Xin Li, Chengfei Jiang, Yang Sun, Xuefeng Wu, Xudong Wu, Huiming Ge, Yan Shen, Qiang Xu
ABSTRACT
Breast cancer is a heterogeneous disease that includes different molecular subtypes. The basal-like subtype has a poor prognosis and a high recurrence rate, whereas the luminal-like subtype confers a more favorable patient prognosis partially due to anti-hormone therapy responsiveness. Here, we demonstrate that diptoindonesin G (Dip G), a natural product, exhibits robust differentiation-inducing activity in basal-like breast cancer cell lines and animal models. Specifically, Dip G treatment caused a partial transcriptome shift from basal to luminal gene expression signatures and prompted sensitization of basal-like breast tumors to tamoxifen therapy. Dip G upregulated the expression of both GABARAPL1 (GABAA receptor-associated protein-like 1) and ERβ. We revealed a previously unappreciated role of GABARAPL1 as a regulator in the specification of breast cancer subtypes that is dependent on ERβ levels. Our findings shed light on new therapeutic opportunities for basal-like breast cancer via a phenotype switch and indicate that Dip G may serve as a leading compound for the therapy of basal-like breast cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human GABARAPL1