MilliporeSigma
  • New strategy to control cell migration and metastasis regulated by CCN2/CTGF.
New strategy to control cell migration and metastasis regulated by CCN2/CTGF.
Cancer cell international (2014-08-15)
Diego Pinheiro Aguiar, Gabriel Correa de Farias, Eduardo Branco de Sousa, Juliana de Mattos Coelho-Aguiar, Julie Calixto Lobo, Priscila Ladeira Casado, Maria Eugênia Leite Duarte, José Garcia Ribeiro Abreu
ABSTRACT
Connective tissue growth factor (CTGF)/CCN family member 2 (CCN2) is a CCN family member of matricellular signaling modulators. It has been shown that CCN2/CTGF mediates cell adhesion, aggregation and migration in a large variety of cell types, including vascular endothelial cells, fibroblasts, epithelial cells, aortic smooth muscle and also pluripotent stem cells. Others matricellular proteins are capable of interacting with CCN2/CTGF to mediate its function. Cell migration is a key feature for tumor cell invasion and metastasis. CCN2/CTGF seems to be a prognostic marker for cancer. In addition, here we intend to discuss recent discoveries and a new strategy to develop therapies against CCN2/CTGF, in order to treat cancer metastasis.