MilliporeSigma
A Requirement for Argonaute 4 in Mammalian Antiviral Defense.
Cell reports (2020-02-13)
Fatemeh Adiliaghdam, Megha Basavappa, Tahnee L Saunders, Dewi Harjanto, John T Prior, D Alexander Cronkite, Nina Papavasiliou, Kate L Jeffrey
ABSTRACT
While interferon (IFN) responses are critical for mammalian antiviral defense, induction of antiviral RNA interference (RNAi) is evident. To date, individual functions of the mammalian RNAi and micro RNA (miRNA) effector proteins Argonautes 1-4 (AGO1-AGO4) during virus infection remain undetermined. AGO2 was recently implicated in mammalian antiviral defense, so we examined antiviral activity of AGO1, AGO3, or AGO4 in IFN-competent immune cells. Only AGO4-deficient cells are hyper-susceptible to virus infection. AGO4 antiviral function is both IFN dependent and IFN independent, since AGO4 promotes IFN but also maintains antiviral capacity following prevention of IFN signaling or production. We identified AGO-loaded virus-derived short interfering RNAs (vsiRNAs), a molecular marker of antiviral RNAi, in macrophages infected with influenza or influenza lacking the IFN and RNAi suppressor NS1, which are uniquely diminished without AGO4. Importantly, AGO4-deficient influenza-infected mice have significantly higher burden and viral titers in vivo. Together, our data assign an essential role for AGO4 in mammalian antiviral defense.
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse