MilliporeSigma
  • Exogenous pulmonary surfactant prevents the development of intra-abdominal adhesions in rats.
Exogenous pulmonary surfactant prevents the development of intra-abdominal adhesions in rats.
Journal of cellular and molecular medicine (2016-02-02)
Alberto Schanaider, Ricardo Cotta-Pereira, Paulo C Silva, Hugo Macedo-Ramos, Johnatas D Silva, Pedro A C Teixeira, Vera L N Pannain, Patricia R M Rocco, Wagner Baetas-da-Cruz
ABSTRACT
Intra-abdominal adhesions are major post-operative complications for which no effective means of prevention is available. We aimed to evaluate the efficacy of exogenous pulmonary surfactant administration in the prevention of post-operative abdominal adhesions. Rats were randomly assigned to undergo laparotomy (L) or gastroenterostomy (GE) and then treated with surfactant (groups L-S and GE-S, respectively). Intra-abdominal adhesions, collagen fibre content, metalloproteinase (MMP)-9, expression of growth factors (TGF-β, KGF and VEGF), type III procollagen (PCIII) and pro-caspase 3, as well as isolectin B4 and ED1-positive cells expressing MMP-9, were evaluated. Groups treated with surfactant (GE-S and L-S) exhibited fewer adhesions. A significant reduction in collagen fibre content was observed in GE-S compared to GE animals (P < 0.001). In situ and gelatin zymography analysis showed higher MMP-9 expression and activity in the GE-S group compared to the GE group (P < 0.05). ED1-positive cell counts were significantly higher in the GE-S group (P < 0.001) than in the GE group. Virtually all cells positive for ED1 were MMP-9+. Double-labelling of MMP-9 with IB4 showed no significant differences between GE-S and GE groups. TGF-β, KGF, PCIII and pro-caspase-3 mRNA expression decreased significantly in GE-S compared to GE animals (P < 0.05). Surfactant administration also reduced apoptosis in the GE-S group. These findings suggest that surfactant reduces the intra-abdominal adhesions triggered by laparotomy and gastrointestinal anastomosis, thus preventing fibrosis formation at the peritoneal surfaces. This preclinical study suggests an innovative treatment strategy for intra-abdominal adhesions with surfactant and to endorse its putative mechanism of action.
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, HRP conjugate, Chemicon®, from rabbit