MilliporeSigma
  • DNA Damage Response Regulates Initiation of Liver Regeneration Following Acetaminophen Overdose.
DNA Damage Response Regulates Initiation of Liver Regeneration Following Acetaminophen Overdose.
Gene expression (2018-03-16)
Prachi Borude, Bharat Bhushan, Udayan Apte
ABSTRACT
Acetaminophen (APAP) overdose is the leading cause of acute liver failure (ALF) with limited treatment options. It is known that liver regeneration following APAP-induced ALF is a deciding factor in the final outcome. Previous studies from our laboratory using an incremental dose model involving a regenerating (300 mg/kg, APAP300) and a nonregenerating (600 mg/kg, APAP600) dose of APAP in mice have revealed several proregenerative pathways that regulate regeneration after APAP overdose. Here we report that DNA damage and repair mechanisms regulate initiation of liver regeneration following APAP overdose. Mice treated with nonregenerating APAP600 dose showed prolonged expression of pH2AX, a marker of the DNA double-strand break (DSB), compared with APAP300. In regenerating APAP300 dose-treated mice, H2AX was rapidly dephosphorylated at Tyr142, indicating timely DNA repair. Expression of several DNA repair proteins was substantially lower with APAP600. Poly(ADP) ribose polymerase (PARP) activation, involved in DNA repair, was significantly higher in the APAP300 group compared to the APAP600 group. Activation of p53, the major cell cycle checkpoint protein, was significantly higher with APAP600 as demonstrated by substantially higher expression of its target genes. Taken together, these data show that massive DNA DSB occurs in high-dose APAP toxicity, and lack of prompt DSB repair after APAP overdose leads to prolonged growth arrest and proliferative senescence, resulting in inhibited liver regeneration.