MilliporeSigma
  • Chickpea (Cicer arietinum) proteins induce allergic responses in nasobronchial allergic patients and BALB/c mice.
Chickpea (Cicer arietinum) proteins induce allergic responses in nasobronchial allergic patients and BALB/c mice.
Toxicology letters (2012-01-31)
Alok Kumar Verma, Sandeep Kumar, Anurag Tripathi, Bhushan P Chaudhari, Mukul Das, Premendra D Dwivedi
ABSTRACT
Allergy to chickpea or Garbanzo bean (Cicer arietinum) has been reported in the Indian population. Little information is found regarding allergenic events involved in the chickpea allergy; therefore, chickpea allergenicity assessment was undertaken. In vivo and ex vivo studies were carried out using BALB/c mice. Chickpea skin prick test positive patients have been used to extend this study in humans. Identification of allergens was carried out by simulated gastric fluids assay for pepsin resistant polypeptides and validated by IgE western blotting using chickpea sensitive humans and sensitized mice sera. Our data have shown the occurrence of a systemic anaphylactic reaction resulting in reduced body temperature after challenge along with significantly increased levels of IgE, IgG1, MMCP-1, CCL-2 as well as histamine. Further, increased Th1/Th2 (mixed) cytokine response was observed in spleen cell culture supernatants. Jejunum, lungs and spleen showed prominent histopathological changes specific for allergic inflammation. Immunoblotting with pooled sera of either sensitized mice or human sera recognized seven similar IgE binding polypeptides that may be responsible for chickpea induced hypersensitivity reactions. This study has addressed the allergenic manifestations associated with chickpea consumption and identifies the proteins responsible for allergenicity which may prove useful in diagnosis and management of allergenicity of legumes especially chickpea.