MilliporeSigma
  • Uniformly cationized protein efficiently reaches the cytosol of mammalian cells.
Uniformly cationized protein efficiently reaches the cytosol of mammalian cells.
Bioconjugate chemistry (2012-09-20)
Midori Futami, Yasuyoshi Watanabe, Takashi Asama, Hitoshi Murata, Hiroko Tada, Megumi Kosaka, Hidenori Yamada, Junichiro Futami
ABSTRACT
Protein cationization techniques are powerful protein transduction methods for mammalian cells. As we demonstrated previously, cationized proteins with limited conjugation to polyethylenimine have excellent ability to enter into cells by adsorption-mediated endocytosis [Futami, J., et al. (2005) J. Biosci. Bioeng. 99, 95-103]. In this study, we show that proteins with extensive and uniform cationization covering the protein surface reach the cytoplasm and nucleus more effectively than proteins with limited cationic polymers or proteins that are fused to cationic peptides. Although extensive modification of carboxylates results in loss of protein function, chicken avidin retains biotin-binding ability even after extensive amidation of carboxylates. Using this cationized avidin carrier system, the protein transduction ability of variously cationized avidins was investigated using biotinylated protein as a probe. The results revealed that cationized avidins bind rapidly to the cell surface followed by endocytotic uptake. Small amounts of uniformly cationized avidin showed direct penetration into the cytoplasm within a 15 min incubation. This penetration route seemed to be energy dependent and functioned under cellular physiological conditions. A biotinylated exogenous transcription factor protein that penetrated cells was demonstrated to induce target gene expression in living cells.
Sigma-Aldrich
3-[2-N-(Biotinyl)aminoethyldithio]propanoic acid, ≥95%