MilliporeSigma
  • Nucleolar stress regulation of endometrial receptivity in mouse models and human cell lines.
Nucleolar stress regulation of endometrial receptivity in mouse models and human cell lines.
Cell death & disease (2019-11-07)
Wei Hu, Yu-Xiang Liang, Jia-Mei Luo, Xiao-Wei Gu, Zi-Cong Chen, Tao Fu, Yu-Yuan Zhu, Shuai Lin, Hong-Lu Diao, Bo Jia, Zeng-Ming Yang
ABSTRACT
Embryo implantation is essential to the successful establishment of pregnancy. A previous study has demonstrated that actinomycin D (ActD) could initiate the activation of mouse delayed implantation. However, the mechanism underlying this activation remains to be elucidated. A low dose of ActD is an inducer of nucleolar stress. This study was to examine whether nucleolar stress is involved in embryo implantation. We showed that nucleolar stress occurred when delayed implantation was activated by ActD in mice. ActD treatment also stimulated the Lif-STAT3 pathway. During early pregnancy, nucleolar stress was detected in the luminal epithelial cells during the receptive phase. Blastocyst-derived lactate could induce nucleolar stress in cultured luminal epithelial cells. The inhibition of nucleophosmin1 (NPM1), which was a marker of nucleolar stress, compromised uterine receptivity and decreased the implantation rates in pregnant mice. To translate these mouse data into humans, we examined nucleolar stress in human endometrium. Our data demonstrated that ActD-induced nucleolar stress had positive effects on the embryo attachment by upregulating IL32 expression in non-receptive epithelial cells rather than receptive epithelial cells. Our data should be the first to demonstrate that nucleolar stress is present during early pregnancy and is able to induce embryo implantation in both mice and humans.
Product Description
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 10.37 mPa.s (25 °C)