MilliporeSigma
  • Abundance of adiponectin mRNA transcript in the buffalo corpus luteum during the estrous cycle and effects on progesterone secretion in vitro.
Abundance of adiponectin mRNA transcript in the buffalo corpus luteum during the estrous cycle and effects on progesterone secretion in vitro.
Animal reproduction science (2019-08-14)
Mahesh Gupta, Ankita Thakre, Krushna Bibichand Bahiram, Vikas Muralidhar Sardar, Shashank Deorao Dudhe, Jayant Pandurang Korde, Sachin Wasudeorao Bonde, Nitin Vasantrao Kurkure
ABSTRACT
Adiponectin is an adipocyte derived cytokine implicated in energy homeostasis, insulin resistance and is involved in the regulation of reproduction both centrally and peripherally in animals. The present study was conducted to investigate adiponectin (ADIPOQ) and its receptors ADIPOR1 and ADIPOR2 abundance of mRNA transcript and protein in different stages of corpora lutea (CL) development during the estrous cycle of water buffalo and to determine the effect of adiponectin on cultured luteal cells of water buffalo (Bubalus bubalis). The results indicate adiponectin, ADIPOR1, and ADIPOR2 were present in buffalo corpora lutea (CL) throughout the estrous cycle. The abundance of adiponectin and its receptors was greater in the early and regressing and was less in mid- and late-stages of CL functionality. Adiponectin and its receptors were localized in the cytoplasm of small and large luteal cells. Furthermore, luteal cells were cultured in the in-vitro culture system and were treated with 1 and 10 μg/mL dose of adiponectin for 48 h. Adiponectin at both doses decreased (P < 0.05) progesterone (P4) secretion from cultured luteal cells and also suppressed the abundance of factors involved in P4productionv [Steroidogenic Acute Regulatory Protein (STAR), cytochrome P45011A1 (CYP11A1) and 3β-hydroxysteroid dehydrogenase (HSD3B1) at the 10 μg/mL dose as compared to adiponectin non-supplemented cells]. In conclusion, results of the present study indicate adiponectin and its receptors are present in bubaline CL and adiponectin inhibits P4 production in cultured luteal cells. The findings indicate adiponectin affects luteal dynamics and reproductive functions in water buffalo.