MilliporeSigma
  • Complexes Formed by Hydrophobic Interaction between Ag-Nanospheres and Adsorbents for the Detection of Methyl Salicylate VOC.
Complexes Formed by Hydrophobic Interaction between Ag-Nanospheres and Adsorbents for the Detection of Methyl Salicylate VOC.
Nanomaterials (Basel, Switzerland) (2019-11-17)
Jinhyuk Park, J Alex Thomasson, Sandun Fernando, Kyung-Min Lee, Timothy J Herrman
ABSTRACT
Surface-enhanced Raman spectroscopy (SERS) has been widely investigated in many applications. However, only little work has been done on using SERS for the detection of volatile organic compounds (VOCs), primarily due to the challenges associated with fabricating SERS substrates with sufficient hotspots for signal enhancement and with the surface interfacially compatible for the VOCs. This study investigated the phase transfer of Ag-nanospheres (AgNSs) from the aqueous phase to the non-aqueous phase by electrostatic interaction induced by cationic surfactants, and the feasibility of the transferred AgNSs as SERS substrates for the determination of methyl salicylate VOC. Results indicated that one of three cationic surfactants, tetraoctylammonium bromide (TOAB) dissolved in organic solvent showed successful phase transfer of the AgNSs confirmed by several characterization analyses. The complex formed by hydrophobic interaction between the transferred AgNSs and Tenax-TA adsorbent polymer was able to be utilized as a SERS substrate, and the volatile of methyl salicylate could be easily determined from SERS measurements at 4 h static volatile collection. Therefore, the proposed new techniques can be effectively employed to areas where many VOCs relevant to food and agriculture need to be analyzed.
Product Description
Supelco
Purge/Trap I Vocarb® 4000, for use with OI Analytical Eclipse 4660, 4560