MilliporeSigma
  • Phenylpiperidine-type γ-secretase modulators target the transmembrane domain 1 of presenilin 1.
Phenylpiperidine-type γ-secretase modulators target the transmembrane domain 1 of presenilin 1.
The EMBO journal (2011-10-18)
Yu Ohki, Takuya Higo, Kengo Uemura, Naoaki Shimada, Satoko Osawa, Oksana Berezovska, Satoshi Yokoshima, Tohru Fukuyama, Taisuke Tomita, Takeshi Iwatsubo
ABSTRACT
Amyloid-β peptide ending at the 42nd residue (Aβ42) is implicated in the pathogenesis of Alzheimer's disease (AD). Small compounds that exhibit selective lowering effects on Aβ42 production are termed γ-secretase modulators (GSMs) and are deemed as promising therapeutic agents against AD, although the molecular target as well as the mechanism of action remains controversial. Here, we show that a phenylpiperidine-type compound GSM-1 directly targets the transmembrane domain (TMD) 1 of presenilin 1 (PS1) by photoaffinity labelling experiments combined with limited digestion. Binding of GSM-1 affected the structure of the initial substrate binding and the catalytic sites of the γ-secretase, thereby decreasing production of Aβ42, possibly by enhancing its conversion to Aβ38. These data indicate an allosteric action of GSM-1 by directly binding to the TMD1 of PS1, pinpointing the target structure of the phenylpiperidine-type GSMs.