MilliporeSigma
  • The impact of porcine circovirus associated diseases on live attenuated classical swine fever vaccine in field farm applications.
The impact of porcine circovirus associated diseases on live attenuated classical swine fever vaccine in field farm applications.
Vaccine (2019-09-11)
Jing-Yuan Chen, Chi-Ming Wu, Chih-Ming Liao, Kun-Chao Chen, Chia-Chin You, Yu-Wen Wang, Chienjin Huang, Maw-Sheng Chien
ABSTRACT
Porcine circovirus associated diseases (PCVADs) are among the most important diseases affecting the worldwide swine industry. Vaccination against porcine circovirus type 2 (PCV2) infection has been utilized for disease control and effectively reduces clinical signs of PCVADs. To evaluate the efficacy of the PCV2 vaccine in field farms, we conducted a trial using conventional pigs immunized with the subunit PCV2 vaccine followed by PCV2 challenge. Immunized pigs demonstrated lower serum viral loads, less viral antigen staining in lymph nodes, and higher average daily weight gain, confirming the protective efficacy of the vaccine. However, low levels of PCV2 infection were still detected in vaccinated pigs after challenge, suggesting that the PCV2 vaccine was unable to eradicate the virus, which could lead to asymptomatic PCV2 subclinical infection (PCV2-SI) in pig farms. Additionally, PCV2 infection is a risk factor for impaired pig immune response development during the weaning to growth stages, which is a crucial period to receive vaccines against classical swine fever (CSF). Therefore, the impact of PCV2-SI or PCV2-systemic disease (PCV2-SD) on live attenuated CSF vaccine was investigated. After PCV2 challenge, there was no difference in levels of classical swine fever virus (CSFV) neutralizing antibodies (NA) between pigs with PCV2-SD and PCV2-SI, suggesting that the efficacy of CSF vaccine was compromised. Moreover, results of long-term monitoring of CSFV NA titers in PCV2-SI pigs with minimized interference by maternally-derived antibodies suggested that serum PCV2 viral loads greater than 102 copies/mL may compromise the efficacy of CSF vaccine. Overall, a conventional pig model was established to demonstrate the impaired efficacy of the subunit PCV2 vaccine and its impact on the CSF vaccine in vaccination-challenge trials. Additionally, the impaired efficacy of the PCV2 vaccine resulted in increased PCV2-SI, eventually leading to compromised the live attenuated CSF vaccine induced NA response in field farm applications.