MilliporeSigma
  • Expression of sarcolectin in the human pituitary gland and amniotic fluid.
Expression of sarcolectin in the human pituitary gland and amniotic fluid.
Biochimie (2010-05-05)
Jean-Pierre M'bika, Wael Khazen, Ammar Achour, Charles Chany
ABSTRACT
Sarcolectin (SCL) is a 55 kDa protein cross-reacting with a cytokeratin 7 monomer found in placental blood, sarcomas and various tissues. It blocks the synthesis of interferon-dependent secondary proteins, induces cell DNA activation and sensitizes cells to viral infection. SCL is a potent promoter of tissue growth. In the present report, we demonstrate that SCL is expressed in the human pituitary gland at the mRNA and protein levels. We show also its presence in human amniotic fluid in high titres while interferon titres is weak. These results allow to postulate a potential role of SCL as a growth factor participating in human foetal development.