MilliporeSigma
  • Enhancement of cellular src gene product associated tyrosyl kinase activity following polyoma virus infection and transformation.
Enhancement of cellular src gene product associated tyrosyl kinase activity following polyoma virus infection and transformation.
Cell (1984-10-01)
J B Bolen, C J Thiele, M A Israel, W Yonemoto, L A Lipsich, J S Brugge
ABSTRACT
We examine the interaction between polyoma-virus-encoded middle tumor antigen and the cellular src gene product, pp60c-src, using a series of monoclonal antibodies that recognize mammalian pp60c-src. Our results show that infection of mouse cells with transformation-competent strains of polyoma virus results in the stimulation of pp60c-src kinase activity severalfold over that observed in uninfected mouse cells and mouse cells infected with transformation-deficient polyoma virus. A similar degree of enhancement of pp60c-src kinase activity was found in polyoma-virus-transformed rodent cells. No differences were detected in the level of pp60c-src synthesis in polyoma-virus-infected and uninfected mouse cells or polyoma-virus-transformed and normal rodent cells. These studies demonstrate that polyoma-virus-encoded middle tumor antigen is associated with pp60c-src in lysates of polyoma-virus-infected and polyoma-virus-transformed cells and suggest a novel mechanism for the functional activation of a cellular proto-oncogene product, namely, that the interaction between middle tumor antigen and pp60c-src leads to a stimulation of pp60c-src tyrosyl kinase activity.