MilliporeSigma
  • Neuron-Astrocyte Metabolic Coupling Protects against Activity-Induced Fatty Acid Toxicity.
Neuron-Astrocyte Metabolic Coupling Protects against Activity-Induced Fatty Acid Toxicity.
Cell (2019-05-28)
Maria S Ioannou, Jesse Jackson, Shu-Hsien Sheu, Chi-Lun Chang, Aubrey V Weigel, Hui Liu, H Amalia Pasolli, C Shan Xu, Song Pang, Doreen Matthies, Harald F Hess, Jennifer Lippincott-Schwartz, Zhe Liu
ABSTRACT
Metabolic coordination between neurons and astrocytes is critical for the health of the brain. However, neuron-astrocyte coupling of lipid metabolism, particularly in response to neural activity, remains largely uncharacterized. Here, we demonstrate that toxic fatty acids (FAs) produced in hyperactive neurons are transferred to astrocytic lipid droplets by ApoE-positive lipid particles. Astrocytes consume the FAs stored in lipid droplets via mitochondrial β-oxidation in response to neuronal activity and turn on a detoxification gene expression program. Our findings reveal that FA metabolism is coupled in neurons and astrocytes to protect neurons from FA toxicity during periods of enhanced activity. This coordinated mechanism for metabolizing FAs could underlie both homeostasis and a variety of disease states of the brain.
Sigma-Aldrich
Anti-Apolipoprotein E Goat pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography