MilliporeSigma
  • Supramolecular Complexes of Graphene Oxide with Porphyrins: An Interplay between Electronic and Magnetic Properties.
Supramolecular Complexes of Graphene Oxide with Porphyrins: An Interplay between Electronic and Magnetic Properties.
Molecules (Basel, Switzerland) (2019-02-17)
Kornelia Lewandowska, Natalia Rosiak, Andrzej Bogucki, Judyta Cielecka-Piontek, Mikołaj Mizera, Waldemar Bednarski, Maciej Suchecki, Konrad Szaciłowski
ABSTRACT
Graphene oxide (GO) was modified by two modified porphyrins (THPP and TCPP) to form GO⁻porphyrin hybrids. Spectroscopic measurements demonstrated the formation of stable supramolecular aggregates when mixing two components in solution. The Fourier transform infrared (FTIR) and Raman scattering measurements confirm π-stacking between hydrophobic regions of GO nanoflakes and porphyrin molecules. On the number and the kind of paramagnetic centers generated in pristine GO samples, which originate from spin anomalies at the edges of aromatic domains within GO nanoflakes. More significant changes in electronic properties have been observed in hybrid materials. This is particularly evident in the drastic increase in the number of unpaired electrons for the THPP-GO sample and the decrease in the number of unpaired electrons for the TCPP-GO. The difference of paramagnetic properties of hybrid materials is a consequence of π-stacking between GO and porphyrin rings. An interesting interplay between modifiers and the surface of GO leads to a significant change in electronic structure and magnetic properties of the designed hybrid materials. Based on the selection of molecular counterpart we can affect the behavior of hybrids upon light irradiation in a different manner, which may be useful for the applications in photovoltaics, optoelectronics, and spintronics.