MilliporeSigma
  • A Histone Acetyltransferase Inhibitor with Antifungal Activity against CTG clade Candida Species.
A Histone Acetyltransferase Inhibitor with Antifungal Activity against CTG clade Candida Species.
Microorganisms (2019-07-18)
Michael Tscherner, Karl Kuchler
ABSTRACT
Candida species represent one of the most frequent causes of hospital-acquired infections in immunocompromised patient cohorts. Due to a very limited set of antifungals available and an increasing prevalence of drug resistance, the discovery of novel antifungal targets is essential. Targeting chromatin modifiers as potential antifungal targets has gained attention recently, mainly due to their role in regulating virulence in Candida species. Here, we describe a novel activity for the histone acetyltransferase inhibitor Cyclopentylidene-[4-(4-chlorophenyl)thiazol-2-yl)hydrazone (CPTH2) as a specific inhibitor of CTG clade Candida species. Furthermore, we show that CPTH2 has fungicidal activity and protects macrophages from Candida-mediated death. Thus, this work could provide a starting point for the development of novel antifungals specific to CTG clade Candida species.