MilliporeSigma
  • Comparison of soybean peroxidase with horseradish peroxidase and alkaline phosphatase used in immunoassays.
Comparison of soybean peroxidase with horseradish peroxidase and alkaline phosphatase used in immunoassays.
Analytical biochemistry (2019-06-16)
Huijuan Yang, Candace S Bever, Huiyan Zhang, Ghulam Mujtaba Mari, Hongfang Li, Xiya Zhang, Liuchuan Guo, Ziwen Wang, Pengjie Luo, Zhanhui Wang
ABSTRACT
Enzyme labeling of an antigen or an antibody helps to visualize and amplify the signal and is an important reagent used in immunoassays for the detection of a target of interest. In this research, soybean peroxidase (SBP), a less commonly used enzyme reporter, was compared in immunoassays with the two most commonly used reagents, horseradish peroxidase (HRP) and alkaline phosphatase (ALP). The enzyme-antibody conjugates were evaluated by their performance in an indirect competitive enzyme-linked immunosorbent assay (icELISA) and in an indirect competitive chemiluminescent enzyme immunoassay (icCLEIA) for ractopamine (RAC). The results revealed that the more affordable SBP offers a long-lasting chemiluminescent signal, which outperformed ALP and HRP. SBP-antibody conjugate (SBP-Ab) based immunoassays produced lower limits of detection (LODs) and better accuracy in the detection of RAC in animal urine samples. Additionally, SBP-Ab has advantages in being more resistant to heat, acid and organic solvents. These results suggest that SBP could be a potentially excellent alternative to HRP and ALP for the development of immunoassay in food safety field.