MilliporeSigma
  • Influence of the amino acid composition on the ionization efficiencies of small peptides.
Influence of the amino acid composition on the ionization efficiencies of small peptides.
Journal of mass spectrometry : JMS (2019-03-09)
Piia Liigand, Karl Kaupmees, Anneli Kruve
ABSTRACT
Electrospray ionization is widely used to generate gas phase ions from a variety of molecules ranging from small ions to large proteins, while the ionization mechanism is claimed to depend on the size of the molecule. For small molecules, the ionization efficiency, the amount of gas phase ions produced in the electrospray process, depends on the properties of the compound. Here, we show that the amino acid composition also influences the ionization efficiency of the oligopeptide. Additionally, we show that the ionization efficiencies of oligopeptides consisting of more than five amino acid residues are very similar to one another, and assuming equal ionization efficiencies is feasible. Therefore, accurate ionization efficiency predictions are needed mainly for small oligopeptides. For these oligopeptides, the amino acid composition can be used to estimate the ionization efficiency in an easy to use manner.