MilliporeSigma
  • MicroRNA-140 inhibits skeletal muscle glycolysis and atrophy in endotoxin-induced sepsis in mice via the WNT signaling pathway.
MicroRNA-140 inhibits skeletal muscle glycolysis and atrophy in endotoxin-induced sepsis in mice via the WNT signaling pathway.
American journal of physiology. Cell physiology (2019-05-03)
Li Liu, Tian-Mei Li, Xue-Ru Liu, Yi-Ping Bai, Jie Li, Ni Tang, Xiao-Bin Wang
ABSTRACT
Sepsis is a systemic inflammatory response syndrome resulting from infection. This study aimed at exploring the role of microRNA-140 (miR-140) in septic mice. Wnt family member 11 (WNT11) was verified to be a target gene of miR-140 after bioinformatic prediction and dual luciferase reporter gene assay. Importantly, miR-140 negatively regulated WNT11. We initially induced the model of sepsis by endotoxin, and then ectopic expression and knockdown experiments were performed to explore the functional role of miR-140 in sepsis. Additionally, cross-sectional areas of muscle fiber, lactic acid production, 3-methylhistidine (3-MH) and tyrosine (Tyr) production in extensor digitorium longus (EDL) muscles, and serum levels of inflammatory factors were examined. The effect of miR-140 on the expression of WNT signaling pathway-related and apoptosis-related factors in skeletal muscle tissue was determined. The experimental results indicated that upregulated miR-140 or silenced WNT11 increased cross-sectional areas of muscle fiber while decreasing lactic acid production, skeletal muscle cell apoptosis [corresponding to downregulated B cell lymphoma 2 (Bcl-2)-associated X protein (Bax) and caspase-3 and upregulated Bcl-2], and the proteolytic rate of Tyr and 3-MH. Also, overexpressed miR-140 or silenced WNT11 reduced inflammation as reflected by decreased serum levels of IL-6, IL-10, and TNF-α. Furthermore, overexpression of miR-140 was shown to suppress the activation of the WNT signaling pathway, accompanied by decreased expression of WNT11, β-catenin, and GSK-3β. Taken together, upregulation of miR-140 could potentially inhibit skeletal muscle lactate release, an indirect measure of glycolysis, and atrophy in septic mice through suppressing the WNT signaling pathway via inhibiting WNT11 expression.