- Impact of Gelatin Supplemented with Gum Arabic, Tween 20, and β-Cyclodextrin on the Microencapsulation of Turkish Oregano Extract.
Impact of Gelatin Supplemented with Gum Arabic, Tween 20, and β-Cyclodextrin on the Microencapsulation of Turkish Oregano Extract.
Microencapsulation protects core materials from deteriorating due to environmental conditions, such as moisture or oxidation, and improves the bioavailability of active compounds, allowing one to make solid formulations from oils and increase their solubility. Wall and core material properties determine the microencapsulation efficiency and the best results are achieved when a wall material mixture is used to prepare the microcapsules. In this work, we optimized the wall material composition (gelatin supplemented with gum Arabic, Tween 20, and β-cyclodextrin) of Turkish oregano microcapsules prepared by spray-drying technology to increase the product yield, the encapsulation efficiency, and to achieve narrower particle size distribution. When the wall material solution contained 10 g of gelatin, 7.5 g of gum Arabic, 1.99 g of Tween 20, 1.98 g of β-cyclodextrin, and 20 g of ethanolic oregano extract, the encapsulation efficiency of oregano's active compounds, rosmarinic acid and carvacrol, were 96.7% and 99.8%, respectively, and the product yield was 85.63%. The physicochemical properties, microscopic morphology, and in vitro release of the prepared microcapsules were characterized in the study. The use of gelatin as the main coating material, in supplementation with gum Arabic, Tween 20, and β-cyclodextrin, not only improved the encapsulation efficiency, but also increased the in vitro release of both main active compounds of Turkish oregano extract-rosmarinic acid and carvacrol.