MilliporeSigma
  • Modulation of nuclear receptor function: Targeting the protein-DNA interface.
Modulation of nuclear receptor function: Targeting the protein-DNA interface.
Molecular and cellular endocrinology (2019-02-01)
Helder Veras Ribeiro Filho, Izabella Luisa Tambones, Marieli Mariano Gonçalves Dias, Natalia Bernardi Videira, Marjorie Bruder, Angélica Amorim Amato, Ana Carolina Migliorini Figueira
ABSTRACT
Nuclear receptors (NRs) are a superfamily of ligand-dependent transcription factors that modulate several biological processes. Traditionally, modulation of NRs has been focused on the development of ligands that recognize and bind to the ligand binding domain (LBD), resulting in activation or repression of transcription through the recruitment of coregulators. However, for more severe diseases, such as breast and prostate cancer, the conventional treatment addressing LBD modulation is not always successful, due to tumor resistance. To overcome these challenges and aiming to modulate NR activity by inhibiting the NR-DNA interaction, new studies focus on the development of molecules targeting alternative sites and domains on NRs. Here, we discuss two different approaches for this alternative NR modulation: one targeting the NR DNA binding domain (DBD); and the other targeting the DNA sites recognized by NRs. Our aim is to present the challenges and perspectives for developing specific inhibitors for each purpose, alongside with already reported examples.