MilliporeSigma
  • Expression of adhesin genes and biofilm formation among Klebsiella oxytoca clinical isolates from patients with antibiotic-associated haemorrhagic colitis.
Expression of adhesin genes and biofilm formation among Klebsiella oxytoca clinical isolates from patients with antibiotic-associated haemorrhagic colitis.
Journal of medical microbiology (2019-05-29)
Abdolmajid Ghasemian, Ashraf Mohabati Mobarez, Shahin Najar Peerayeh, Amin Talebi Bezmin Abadi, Sepideh Khodaparast, Suhad Saad Mahmood
ABSTRACT
Biofilm formation and resistance to last-line antibiotics have restricted chemotherapy options toward infection eradication. Fifty K. oxytoca isolates were collected from patients with antibiotic-associated haemorrhagic colitis (AAHC). Antibiotic susceptibility tests were conducted and phenotypic biofilm formation was assessed using microtitre tissue plate (MTP) assay. PCR was employed to amplify the adhesins, extended-spectrum β-lactamases (ESBLs), carbapenemase and colistin resistance genes. The expression of adhesin genes was evaluated using quantitative real-time PCR (RT-qPCR).Results/Key findings. The previous antibiotic consumption and hospitalization (P<0.05) and older ages (P=0.0033) were significantly associated with AAHC. None of the isolates produced biofilm strongly, but 70% of them produced moderate-level biofilm. The blaCTX-M (12/14), the blaIMP (8/14 MICIMI =4 µg ml-1 ) and blaOXA-48-like (5/14) and mcr-1 (4/14) genes were predominant, three of which harbouring all the genes. The expression of matB (0.023) and mrkA (0.011) was significantly different between multidrug-resistant and susceptible isolates. Furthermore, moderately biofilm producer isolates significantly exhibited higher expression of fimA (P=.0117), pilQ (P=0.002) and mrkA (P=0.020) genes compared to biofilm non-producers. No significant difference regarding gene expression was observed among ESBL alleles. Bacterial attachment by adhesins and biofilm formation among extensive drug-resistant K. oxytoca isolates hinder the efficient infection eradication. Hence, control and surveillance studies should be performed and other therapeutic auspicious approaches must be taken into account against AAHC, biofilm formation and drug resistance spread. Furthermore, previous antibiotic consumption and long-term hospitalization should be controlled.