MilliporeSigma
  • Fabrication of Troponin I Biosensor Composed of Multi-Functional DNA Structure/Au Nanocrystal Using Electrochemical and Localized Surface Plasmon Resonance Dual-Detection Method.
Fabrication of Troponin I Biosensor Composed of Multi-Functional DNA Structure/Au Nanocrystal Using Electrochemical and Localized Surface Plasmon Resonance Dual-Detection Method.
Nanomaterials (Basel, Switzerland) (2019-08-03)
Taek Lee, Jinmyeong Kim, Inho Nam, Yeonju Lee, Ha Eun Kim, Hiesang Sohn, Seong-Eun Kim, Jinho Yoon, Sang Woo Seo, Min-Ho Lee, Chulhwan Park
ABSTRACT
In the present study, we fabricated a dual-mode cardiac troponin I (cTnI) biosensor comprised of multi-functional DNA (MF-DNA) on Au nanocrystal (AuNC) using an electrochemical method (EC) and a localized surface plasmon resonance (LSPR) method. To construct a cTnI bioprobe, a DNA 3 way-junction (3WJ) was prepared to introduce multi-functionality. Each DNA 3WJ arm was modified to possess a recognition region (Troponin I detection aptamer), an EC-LSPR signal generation region (methylene blue: MB), and an anchoring region (Thiol group), respectively. After an annealing step, the multi-functional DNA 3WJ was assembled, and its configuration was confirmed by Native-TBM PAGE for subsequent use in biosensor construction. cTnI was also expressed and purified for use in biosensor experiments. To construct an EC-LSPR dual-mode biosensor, AuNCs were prepared on an indium-tin-oxide (ITO) substrate using an electrodeposition method. The prepared multi-functional (MF)-DNA was then immobilized onto AuNCs by covalent bonding. Field emission scanning electron microscope (FE-SEM) and atomic force microscopy (AFM) were used to analyze the surface morphology. LSPR and electrochemical impedance spectroscopy (EIS) experiments were performed to confirm the binding between the target and the bioprobe. The results indicated that cTnI could be effectively detected in the buffer solution and in diluted-human serum. Based on the results of these experiments, the loss on drying (LOD) was determined to be 1.0 pM in HEPES solution and 1.0 pM in 10% diluted human serum. Additionally, the selectivity assay was successfully tested using a number of different proteins. Taken together, the results of our study indicate that the proposed dual-mode biosensor is applicable for use in field-ready cTnI diagnosis systems for emergency situations.