MilliporeSigma
  • Lower rate of early pregnancy loss in patients experiencing early-onset low LH in GnRH antagonist cycles supplemented with menotropin.
Lower rate of early pregnancy loss in patients experiencing early-onset low LH in GnRH antagonist cycles supplemented with menotropin.
Journal of the Formosan Medical Association = Taiwan yi zhi (2018-02-07)
Po-Kai Yang, Ming-Yih Wu, Kuang-Han Chao, Chin-Hao Chang, Mei-Jou Chen, Shee-Uan Chen
ABSTRACT
The role of LH during controlled ovarian stimulation (COS) in the general population remains contentious. There is no consensus on the indications for LH supplementation during COS. The purpose of this study is to determine whether menotropin supplement is associated with decreases in early pregnancy loss rates in patients exhibiting low endogenous LH during COS. This is a single-center, retrospective cohort from a university-affiliated hospital. Patients were enrolled from the in-vitro fertilization center from January, 2011 to December, 2014. Patients who experienced a LH level ≦ 0.8 mIU/mL during stimulation were identified, and patients that received menotropin supplementation were compared to those without menotropin supplementation. Outcome variables, including the number of oocytes retrieved, embryos obtained, implantation rates, pregnancy rates and early pregnancy loss rates, were compared. Patients that experienced low LH during GnRH antagonist protocol and were supplemented with menotropin were associated with lower early pregnancy loss when compared with patients without menotropin supplementation (26.7% vs. 11.5%, p = 0.045). More specifically, in patients who exhibited early-onset low LH, before the use of GnRH antagonists, menotropin supplementation was associated with significantly lower early pregnancy loss compared with non-supplemented patients (3.3% vs. 29.0%, OR: 0.08, p = 0.012). Beneficial effects persisted after adjusting for confounders (aOR: 0.103, 95% CI: 0.011-0.933). Menotropin supplementation is associated with decreased early pregnancy loss in patient who exhibited low LH during GnRH antagonist cycles. This effect is especially prominent in patients who experience low LH before the start of GnRH antagonists.